Orders LinkContactUsLinkComponent
Crystallizing Dish
Crystallizing Dish
Supplier:   Wards
Crystallizing Dish
Supplier:  Wards
Certificates
Crystallizing Dish
Crystallizing Dish
4 Options Available Below
Product Details & Documents
Ideal for storage and crystallization.
  • Borosilicate glass
  • Flat bottom
  • Easy pour spout
Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.
Product Family Options
Product Information
  • Capacity
    Ø
    Height
    Availability
    Price
  • 40 ml
    50 mm
    30 mm
    Log in for availability
    Log in for availability
    Each
    Each - $11.55
    $11.55
    Each
       $11.55
    Supplier Number: CH0066D
  • 100 ml
    70 mm
    40 mm
    Log in for availability
    Log in for availability
    Each
    Each - $15.95
    $15.95
    Each
       $15.95
    Supplier Number: CH0066E
  • 300 ml
    100 mm
    50 mm
    Discontinued
    Supplier Number: CH0066A
  • 1000 ml
    150 mm
    75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $59.90
    $59.90
    Each
       $59.90
    Supplier Number: CH0066B
  • 2000 ml
    190 mm
    100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $103.00
    $103.00
    Each
       $103.00
    Supplier Number: CH0066C