Accredo Safety Push Button Lockout-Electrical Panels, ZING Enterprises
Catalog # 76026-344
Supplier: ZING Enterprises
Specifications
- Description:Normal Push Button Lockout for Electrical Panels
- Length:4"
- Material:Plastic
- Cat. no.:76026-344
- Width:4"
Specifications
About this item
Supports OSHA Standard 1910.147.
- Durable Plastic (Polyurethane) Construction
- High quality
- Safety device
Push Button Lockout provides a safe and effective method for locking out emergency push buttons. The lockout is made from durable plastic (polyurethane) and measures 2.25’’H x 4’’L x 4’’W. The lockout accommodates 1 padlock with a maximum shackle diameter of ¼’’. Includes a label with the legend, “DANGER Do Not Remove.”
Certifications: OSHA 1910.147
Frequently Bought Together