Order Entry
ContactUsLinkComponent
Accredo Safety Electrical Panel Handle Lockout, ZING Enterprises
Accredo Safety Electrical Panel Handle Lockout, ZING Enterprises
Catalog #: 76026-342
Supplier:  ZING Enterprises
LOCKOUT ELECTRICAL PANEL NYLON BASE/ SS
Accredo Safety Electrical Panel Handle Lockout, ZING Enterprises
Catalog #: 76026-342
Supplier:  ZING Enterprises

Specifications

  • Description:
    Electrical panel handle lockout
  • Material:
    Nylon
  • Cat. no.:
    76026-342
  • Length:
    4"
  • Width:
    2.25"

Specifications

About this item

Supports OSHA Standard 1910.147.

  • Nylon base
  • Stainless steel strip
  • High quality

Electrical Panel Handle Lockout prohibits the movement of handles or levers on electrical panels. A yellow nylon base and a flexible stainless steel strip (1'' x 11 4/5'') with 20 holes
makes it impossible for the handles to move from their position once locked with a lockout padlock.
This lockout is recommended for all handles and levers that do not have a built-in lockout arrangement. Accommodates 1 padlock, with shackles up to ¼’’.

Certifications: OSHA 1910.147
Customers Who Bought This Also Bought