Accredo Safety Ball Valve Lockout, ZING Enterprises
Supplier: ZING Enterprises
Certificates
About this item
Convenient way to lockout ball valves.
- High quality
- Effective lockout device
- Very durable
Small Ball Valve Lockout, is made from non-conductive, durable plastic (polyurethane), and fits pipe sizes 3/8" to 1-1/4", and handle sizes 3/8" to 1-1/4".
Ball Valve Lockout, is made from non-conductive, durable plastic (polyurethane), and fits pipe sizes 1-1/2" to 2-1/2", and handle sizes 1-1/2" to 2-1/2".
Large Ball Valve Lockout, is made from non-conductive, durable plastic (polyurethane), and fits pipe sizes 2" to 8", and handle sizes 2" to 8".
Certifications: OSHA 1910.147