EZBio® Vent Filter, 0.22 µm, Nonsterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
Certificates
About this item
EZBio® PTFE vent filters are used with EZBio® Single-Use bottle assemblies, which are perfect for fluid transfer and storage.
- Vent filters used with EZBio® single-use bottle assemblies
- PTFE membrane
- Polypropylene housing
- 0.22 µm
Certifications: All materials comply with USP Class VI standards.
Customers Who Bought This Also Bought