Avantor® Prevail Cyano, HPLC Columns
Supplier: Avantor
Certificates
About this item
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
- Stable from highly organic to highly aqueous
- Specialty phases for specific applications
- Excellent sensitivity with microbore and ELSD applications
- USP L10
General purpose cyano suitable for normal or reversed phase use. For rugged normal phase applications
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
Accessories information: Use guard cartridges with holder (80101/N) and coupler (HI-081).
Specifications
- Catalog No:
- 75797-814
- 75797-812
- 75830-698
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)