Terrific Broth Complete with 100 µg/ml Carbenicillin Bottled Media, Teknova
Catalog # CA100219-278
Supplier: Teknova
CAS Number:
Specifications
- Description:Terrific Broth Complete with 100 µg/mL Carbenicillin
- Cat. no.:CA100219-278
- Volume:1 L
- Supplier No.:T7030
Specifications
About this item
Used to obtain high yields of plasmid DNA from E.coli cultures.
- 1 Liter
- Sterile
Suitable for use with carbenicillin (ampicillin) resistant strains and strains harboring plasmids such as pBluescript, pGEM, and pUC. Contains 1.2% Tryptone, 2.4% yeast extract, 0.4% glycerol, 72mM K2HPO4, and 17mM KH2PO4 with 100µg/ml carbenicillin. Ship on ice. Store frozen.