Orders LinkContactUsLinkComponent
Dissecting Scissors, Sklar
Dissecting Scissors, Sklar
Supplier:   Sklar
Dissecting Scissors, Sklar
Supplier:  Sklar
Certificates
Dissecting Scissors, Sklar
Dissecting Scissors, Sklar
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Sklar instruments are industry standard.
  • Reusable
  • Nonsterile
Product Family Options
Product Information
  • Length
    Material
    Blade type
    Curvature
    Availability
    Price
  • 75995-950
    Dissecting Scissors, Flat Handle with Thumb Spring
    Dissecting Scissors, Flat Handle with Thumb Spring
    150 mm (6")
    Stainless steel
    Blunt/Blunt
    Straight
    Discontinued
    Supplier Number: -
  • 75995-952
    Dissecting Scissors, Round Handle
    Dissecting Scissors, Round Handle
    150 mm (6")
    Stainless steel
    Blunt/Blunt
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $2,110.37
    $2,110.37
    New Customer Promotional Price
    Each
       $2,110.37
    Supplier Number: 98-7803
  • 75995-954
    Dissecting Scissors, Round Handle
    Dissecting Scissors, Round Handle
    180 mm (7")
    Stainless steel
    Blunt/Blunt
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $2,175.83
    $2,175.83
    New Customer Promotional Price
    Each
       $2,175.83
    Supplier Number: 98-7804
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)