If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Length:241 mm (9½")
- Tip Configuration:Fenestrated
- Curvature:Straight
- Cat. no.:75995-394
Merit™ Sterile Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis.
- Securely grips gauze and holds it in place
- Ready-to-use
- Office grade
The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. Merit™ Sterile instruments are ready-to-use, sterile, disposable, office-grade instruments most commonly utilized for routine and in-office minor procedures. They are sold in bulk quantities.