ContactUsLinkComponent
BD BBL™ Stacker™ I Plate™ Trypticase™ Soy Agar, with 5% Sheep Blood/MacConkey II Agar, BD Diagnostics
BD BBL™ Stacker™ I Plate™ Trypticase™ Soy Agar, with 5% Sheep Blood/MacConkey II Agar, BD Diagnostics
  CA-90006-224
 :  BD
 :  
BD BBL™ Stacker™ I Plate™ Trypticase™ Soy Agar, with 5% Sheep Blood/MacConkey II Agar, BD Diagnostics
  CA-90006-224
 :  BD
 :  221291
 :  
  • Description:
    Trypticase™ Soy Agar, with 5% Sheep Blood (TSA II) / MacConkey II Agar
  • Cat. no.:
    CA-90006-224
  • Supplier No.:
    221291
Prepared plated media are designed for the isolation of microorganisms from samples or specimens.
  • Two-section plates containing one or two media formulations
  • Interlocking feature minimizes hazard of sliding plates
  • Ready for immediate use
  • Stackable
These 100x15mm plates feature two sections containing one or two media formulations. Plates will interlock, minimizing the hazard of sliding plates. Plates are ready for immediate use. Store at 2 to 8°C (36 to 46°F). Stackable.