Luer Lock Valve Adaptors, Restek
10862-648
: Restek
- Description:Luer Lock Valve Adaptor for Syringes
- Cat. no.:10862-648
- Supplier No.:21283
Use a luer lock valve adaptor to connect a luer lock fitting to a Rheodyne or Valco valve.
Use a luer lock valve adaptor to connect a luer lock fitting to a Rheodyne or Valco valve.