Force EXP Guard Column Cartridges, Restek
: Restek
Free-Turn architecture lets you change cartridges by hand without breaking inlet/outlet fluid connections, with no tools needed.
- Titanium hybrid ferrules
- High pressure seal
- Auto-adjusting design
Patented titanium hybrid ferrules can be installed repeatedly without compromising high-pressure seal. Auto-adjusting design provides ZDV (zero dead volume) connection to any 10-32 female port. Guard column cartridges require EXP direct connect holder. Pair with EXP hand-tight fitting for tool-free installation. For use with 3 or 5 µm Force LC columns. For 1.8 µm Force columns, use a 0.2 µm UltraShield Filter.
: EXP Direct Connect Holder is available separately.
: To help protect your investment and further extend the life of our already-rugged LC columns, Restek offers the patent-pending guard column hardware developed by Optimize Technologies. A Restek LC guard cartridge in an EXP direct connect holder is the ultimate in column protection.
- Catalog No:
- 75854-232
- 75854-228
- 75854-230
- 75854-284
- 75854-178
- 75854-180
- 75854-176
- 75854-280
- 75854-282
- Max. pressure:
- 600 bar/8,700 psi
- 600 bar/8,700 psi
- 600 bar/8,700 psi
- 600 bar/8,700 psi
- 600 bar/8,700 psi
- 600 bar/8,700 psi
- 600 bar/8,700 psi
- 600 bar/8,700 psi
- 600 bar/8,700 psi
Frequently Bought Together