ContactUsLinkComponent
MIR Performance Validation Kit, PerkinElmer
MIR Performance Validation Kit, PerkinElmer
  10133-738
MIR Performance Validation Kit, PerkinElmer
  10133-738
 :  L1250404
  • Description:
    Mid Infrared IPV kit for Spectrum 10 Software
  • Thickness:
  • Cat. no.:
    10133-738
  • Supplier No.:
    L1250404
Instrument performance validation kits are available for both Mid-IR and Near-IR FT-IR instruments
Upgrade kits are available to replace the polystyrene TRM once the validity period has expired.
 : Kits include a polystyrene Traceable Reference Material (TRM) for abscissa calibration valid for three years, NG11 Schott Glass Reference Material (RM).