Orders LinkContactUsLinkComponent
Decapper for Vials, MicroSlov
Decapper for Vials, MicroSlov
Decapper Tool, 20 mm
Catalog # 97054-996
Supplier #:  95025-DC
Decapper for Vials, MicroSlov
Catalog # 97054-996
Decapper for Vials, MicroSlov
Decapper for Vials, MicroSlov
Decapper Tool, 20 mm
Price
$587.52
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
These decapper tools are used for detaching aluminum Headspace Caps.
Specifications
  • Description:
    Decapper Tool
  • Seal size:
    20 mm
  • Cat. No.:
    97054-996