Order Entry
Canada
ContactUsLinkComponent
ORP Test Solution, WTW
ORP Test Solution, WTW
Catalog # 80085-326
Supplier:  WTW
CAS Number:  
ORP Test Solution, WTW
Catalog # 80085-326
Supplier:  WTW
Supplier Number:  109740
CAS Number:  

Specifications

  • Concentration:
    Uh = 427 Mv (Ch)
  • Description:
    ORP Test Solution
  • Size:
    250 ml
  • Cat. no.:
    80085-326
  • Supplier No.:
    109740

Specifications

About this item

ORP test solution for electrolyte cleaning agents to test, service ORP electrodes easy conditioning and cleaning.