Sodium hydroxide 0.1 mol/L in aqueous solution, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Catalog # CA1.37058.0001
Supplier: EMD MILLIPORE # PCS CA EXCEP
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. no.:CA1.37058.0001
- Supplier No.:1.37058.0001
Specifications
Specification Test Results
- Assay (acidimetric,NaOH):0.05- 0.15 mol/l
- Carbonate (as Na2CO3):≤ 0.2%
- Chloride (Cl):≤ 5%
- Sulfate (SO.):≤ 30 ppm
- Heavy metals (as Pb):≤ 5.0 ppm
- Al (Aluminium):≤ 10 ppm
- Fe (Iron):≤ 3 ppm
- Residual solvents (ICH Q3C):excluded by production process
- Endotoxins:≤ 2.0 I.U./ml
Specification Test Results
About this item
:
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,
:Filtered through a 0.2 µm filter.
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548