Specifications
- Material:Stainless Steel
- Cat. no.:75995-376
- Length:140 mm (51/2")
- Blade Type:Smooth, Angled, Blunt/Blunt
Specifications
About this item
Merit™ Lister Sterile Bandage Scissors are a versatile instrument with multiple uses.
- Angled jaws
- Lower blade is slightly longer
- Flattened blunt nodule on tip of lower blade
The scissors are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric, and occasionally umbilical cord. The jaws are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Sterile Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger. Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures.