Order Entry
Canada
ContactUsLinkComponent
FAME #15 Mix (8 components), Restek
FAME #15 Mix (8 components), Restek
Catalog # RK35036
Supplier:  Restek
CAS Number:  
undefined
FAME #15 Mix (8 components), Restek
Catalog # RK35036
Supplier:  Restek
Supplier Number:  35036
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fatty acid methyl ester, FAME Std #15
  • Size:
    50 mg
  • Cat. no.:
    RK35036
  • Supplier No.:
    35036

Specifications

About this item

Contains: Chain, Compound (CAS#), % by Weight; C16:00 Methyl palmitate (112-39-0), 10%; C18:00 Methyl stearate (112-61-8), 3%; C18:01 Methyl oleate (112-62-9), 50%; C18:02 Methyl linoleate (112-63-0), 30%; C20:00 Methyl arachidate (1120-28-1), 1.5%; C20:01 Methyl eicosenoate (20839-34-3), 1.5%; C22:00 Methyl behenate (929-77-1), 3%; C24:00:00 Methyl lignocerate (2442-48-1), 1%