FAME #14 Mix (11 components), Restek
RK35035
: Restek
:
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME Std #14
- Size:50 mg
- Cat. no.:RK35035
- Supplier No.:35035
Contains: Chain, Compound (CAS#), % by Weight; C14:00 Methyl myristate (124-10-7), 0.1%; C16:00 Methyl palmitate (112-39-0), 26.3%; C16:01 Methyl palmitoleate (1120-25-8), 0.4%; C17:00 Methyl heptadecanoate (1731-92-6), 0.3%; C18:00 Methyl stearate (112-61-8), 33.7%; C18:01 Methyl oleate (112-62-9), 34.3%; C18:02 Methyl linoleate (112-63-0), 3.1%; C18:03 Methyl linolenate (301-00-8), 0.2%; C20:00 Methyl arachidate (1120-28-1), 1.3%; C20:01 Methyl eicosenoate (20839-34-3), 0.1%; C22:00 Methyl behenate (929-77-1), 0.2%