Order Entry
Canada
ContactUsLinkComponent
FAME #20 Mix (8 components), Restek
FAME #20 Mix (8 components), Restek
Catalog # RK35026
Supplier:  Restek
CAS Number:  
undefined
FAME #20 Mix (8 components), Restek
Catalog # RK35026
Supplier:  Restek
Supplier Number:  35026
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fatty acid methyl ester, FAME #20
  • Size:
    50 mg/Ampule
  • Cat. no.:
    RK35026
  • Supplier No.:
    35026

Specifications

About this item

This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like coconut, palm kernel, babassu, and ouri-curi oil.

This mix contains eight components such as C8:00 methyl caprylate (7%), C10:00 methyl caprate (5%), C12:00 methyl laurate (48%), C14:00 methyl myristate (15%), C16:00 methyl palmitate (7%), C18:00 methyl stearate (3%), C18:01 methyl oleate (12%), C18:02 methyl linoleate (3%).

% represents weight/weight% basis (minimum 50 mg/ampoule).