FAME #16 Mix (6 components), Restek
Catalog # RK35022
Supplier: Restek
CAS Number:
Specifications
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME #16
- Size:50 mg
- Cat. no.:RK35022
- Supplier No.:35022
Specifications
About this item
Contains: Chain, Compound (CAS#), % by Weight; C16:00 Methyl palmitate (112-39-0), 6%; C18:00 Methyl stearate (112-61-8), 3%; C18:01 Methyl oleate (112-62-9), 35%; C18:02 Methyl linoleate (112-63-0), 50%; C18:03 Methyl linolenate (301-00-8), 3%; C20:00 Methyl arachidate (1120-28-1), 3%