FAME #8 Mix (5 components), Restek
Catalog # RK35017
Supplier: Restek
CAS Number:
Specifications
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME Std #8
- Size:50 mg
- Cat. no.:RK35017
- Supplier No.:35017
Specifications
About this item
Contains: Chain, Compound (CAS#), % by Weight; C11:00 Methyl undecanoate (1731-86-8), 20%; C12:00 Methyl laurate (111-82-0), 20%; C13:00 Methyl tridecanoate (1731-88-0), 20%; C14:00 Methyl myristate (124-10-7), 20%; C15:00 Methyl pentadecanoate (7132-64-1), 20%