FAME #6 Mix (5 components), Restek
RK35015
: Restek
:
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME Std #6
- Size:50 mg
- Cat. no.:RK35015
- Supplier No.:35015
Contains: Chain, Compound (CAS#), % by Weight; C20:00 Methyl arachidate (1120-28-1), 20%; C20:01 Methyl eicosenoate (20839-34-3), 20%; C20:02 Methyl eicosadienoate (2463-02-7), 20%; C20:03 Methyl homo gamma linolenate (16326-32-2), 20%; C20:04 Methyl arachidonate (2566-89-4), 20%