Order Entry
Canada
ContactUsLinkComponent
FAME #3 Mix (5 components), Restek
FAME #3 Mix (5 components), Restek
Catalog # RK35012
Supplier:  Restek
CAS Number:  
undefined
FAME #3 Mix (5 components), Restek
Catalog # RK35012
Supplier:  Restek
Supplier Number:  35012
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fatty acid methyl ester, FAME Std #3
  • Size:
    50 mg
  • Cat. no.:
    RK35012
  • Supplier No.:
    35012

Specifications

About this item

Contains: Chain, Compound (CAS#), % by Weight; C8:00 Methyl caprylate (111-11-5), 20%; C10:00 Methyl caprate (110-42-9), 20%; C12:00 Methyl laurate (111-82-0), 20%; C14:00 Methyl myristate (124-10-7), 20%; C16:00 Methyl palmitate (112-39-0), 20%