FAME #2 Mix (5 components), Restek
Catalog # RK35011
Supplier: Restek
CAS Number:
Specifications
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME Std #2
- Size:50 mg
- Cat. no.:RK35011
- Supplier No.:35011
Specifications
About this item
Contains: Chain, Compound (CAS#), % by Weight; C6:00 Methyl caproate (106-70-7), 20; C8:00 Methyl caprylate (111-11-5), 20; C10:00 Methyl caprate (110-42-9), 20; C12:00 Methyl laurate (111-82-0), 20; C14:00 Methyl myristate (124-10-7), 20