Herbicide Surrogate (2,4-Dichlorophenyl acetic acid methyl ester (DCAA methyl ester)), Derivatized Form, Restek
Catalog # RK32050
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml in hexane
- CAS No.:55954-23-9
- Description:2,4-Dichlorophenyl acetic acid methyl ester
- Size:1 ml/Ampule
- Formula:C₉H₈Cl₂O₂
- Description:Methyl 2,4-Dichlorophenylacetate
- Cat. no.:RK32050
- Certification:CRM
- Supplier No.:32050
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.