608 Calibration Mix (16 components), Restek
Catalog # RK32022
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in hexane:toluene (1:1)
- Description:608 Calibration Mix
- Size:1 ml/Ampule
- Cat. no.:RK32022
- Certification:CRM
- Supplier No.:32022
Specifications
About this item
This mix contains 16 components.
Contains: Aldrin (309-00-2); α-BHC (319-84-6); β-BHC (319-85-7); δ-BHC (319-86-8); γ-BHC (Lindane) (58-89-9); 4,4'-DDD (72-54-8); 4,4'-DDE (72-55-9); 4,4'-DDT (50-29-3); Dieldrin (60-57-1); Endosulfan I (959-98-8); Endosulfan II (33213-65-9); Endosulfan sulfate (1031-07-8); Endrin (72-20-8); Endrin aldehyde (7421-93-4); Heptachlor (76-44-8); Heptachlor epoxide (isomer B) (1024-57-3)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.