CLP GPC Calibration Mix (5 components), Restek
Catalog # RK32019
Supplier: Restek
CAS Number:
Specifications
- Concentration:In methylene chloride
- Description:CLP GPC Calibration Mix
- Size:1 ml/Ampule
- Cat. no.:RK32019
- Supplier No.:32019
Specifications
About this item
Contains: Bis(2-ethylhexyl) phthalate (117-81-7), 10 mg/mL; Corn oil (8001-30-7), 250 mg/mL; Methoxychlor (72-43-5), 2.0 mg/mL; Perylene (198-55-0), 0.2 mg/mL; Sulfur (7704-34-9), 0.8 mg/mL