Order Entry
Canada
ContactUsLinkComponent
Antifoam Agent for Purge-and-Trap Samples, Restek
Antifoam Agent for Purge-and-Trap Samples, Restek
Catalog # RK31822
Supplier:  Restek
CAS Number:  
undefined
Antifoam Agent for Purge-and-Trap Samples, Restek
Catalog # RK31822
Supplier:  Restek
Supplier Number:  31822
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Antifoam agent for purge-and-trap
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK31822
  • Supplier No.:
    31822

Specifications

About this item

Foam generated as purge gas passes through a sample can enter the analytical trap and possibly the GC column. This silica-containing antifoam agent is effective over a wide pH range and will not conflict with chromatography of target analytes.

  • Efficiently controls foam; effective over a wide pH range.
  • No hazardous materials, no components that are target analytes.
  • Effective at less than 0.1% of sample volume.