Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Appendix IX Mix #2 (32 components), Restek
Appendix IX Mix #2 (32 components), Restek
Appendix IX Mix #2 (32 components), Restek
RK31806
:
Restek
:
Appendix IX Mix #2 (32 components), Restek
RK31806
:
Restek
:
31806
:
Concentration:
1000 µg/ml each in methylene chloride
Description:
Appendix IX Mix #2
Size:
1 ml/Ampule
Cat. no.:
RK31806
Certification:
CRM
Supplier No.:
31806