MA EPH Aliphatic Hydrocarbon Standard (14 components), Restek
RK31459
: Restek
:
- Concentration:1000 µg/ml each in hexane
- Description:MA EPH Aliphatic Hydrocarbon Standard
- Size:1 ml/Ampule
- Cat. no.:RK31459
- Certification:CRM
- Supplier No.:31459
Contains: n-nonane, C9, n-decane, C10, n-dodecane, C12, n-tetradecane, C14, n-hexadecane, C16, n-octadecane, C18, n-nonadecane, C19, n-eicosane, C20, n-docosane, C22, n-tetracosane, C24, n-hexacosane, C26, n-octacosane, C28, n-triacontane, C30, n-hexatriacontane, C36.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.