Specifications
- Description:D2887 Calibration Mix
- Size:1 ml/Ampule
- Cat. no.:RK31222
- Certification:CRM
- Supplier No.:31222
Specifications
About this item
ASTM Methods D2887 and D3710-95
These calibration mixtures are made with pure, highly characterized neat material and are prepared using NIST-traceable balances and weights. Each ampul is supplied with a data sheet indicating the exact concentration and a sample chromatogram.
Contains: Compound, Conc.; (C6) n-Hexane (110-54-3), 6% w/w; (C7) n-Heptane (142-82-5), 6% w/w; (C8) n-Octane (111-65-9), 8% w/w; (C9) n-Nonane (111-84-2), 8% w/w; (C10) n-Decane (124-18-5), 12% w/w; (C11) n-Undecane (1120-21-4), 12% w/w; (C12) n-Dodecane (112-40-3), 12% w/w; (C14) n-Tetradecane (629-59-4), 12% w/w; (C16) n-Hexadecane (544-76-3), 10% w/w; (C18) n-Octadecane (593-45-3), 5% w/w; (C20) n-Eicosane (112-95-8), 2% w/w; (C24) n-Tetracosane (646-31-1), 2% w/w; (C28) n-Octacosane (630-02-4), 1% w/w; (C32) n-Dotriacontane (544-85-4), 1% w/w; (C36) n-Hexatriacontane (630-06-8), 1% w/w; (C40) n-Tetracontane (4181-95-7), 1% w/w; (C44) n-Tetratetracontane (7098-22-8), 1% w/w