607 Nitrosamines Calibration Mix (3 components), Restek
Catalog # RK31032
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methanol
- Description:607 Nitrosamines Calibration Mix
- Size:1 ml/Ampule
- Cat. no.:RK31032
- Certification:CRM
- Supplier No.:31032
Specifications
About this item
Contains: N-Nitrosodimethylamine (62-75-9); N-Nitroso-di-n-propylamine (621-64-7); N-Nitrosodiphenylamine (86-30-6)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.