606 Phthalate Esters Calibration Mix (6 components), Restek
Catalog # RK31031
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methanol
- Description:606 Phthalate Esters Calibration Mix
- Size:1 ml/Ampule
- Cat. no.:RK31031
- Certification:CRM
- Supplier No.:31031
Specifications
About this item
Contains: Bis(2-ethylhexyl)phthalate (117-81-7); Benzyl butyl phthalate (85-68-7); Di-n-butyl phthalate (84-74-2); Diethylphthalate (84-66-2); Dimethylphthalate (131-11-3); Di-n-octyl phthalate (117-84-0)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.