Acid Matrix Spike Mix (5 components), Restek
Catalog # RK31014
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methanol
- Description:Acid Matrix Spike Mix
- Size:1 ml/Ampule
- Cat. no.:RK31014
- Certification:CRM
- Supplier No.:31014
Specifications
About this item
Contains: 4-Chloro-3-methylphenol (59-50-7); 2-Chlorophenol (95-57-8); 4-Nitrophenol (100-02-7); Pentachlorophenol (87-86-5); Phenol (108-95-2)
- Highest concentrations commercially available.
- Reduces laboratory cost per sample extract.