Order Entry
Canada
ContactUsLinkComponent
SV Calibration Mix #6 (18 Components), Restek
SV Calibration Mix #6 (18 Components), Restek
Catalog # RK31012
Supplier:  Restek
CAS Number:  
undefined
SV Calibration Mix #6 (18 Components), Restek
Catalog # RK31012
Supplier:  Restek
Supplier Number:  31012
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in toluene:hexane (1:1)
  • Description:
    SV Calibration Mix #6
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK31012
  • Certification:
    CRM
  • Supplier No.:
    31012

Specifications

About this item

Contains: Aldrin (309-00-2); α-BHC (319-84-6); β-BHC (319-85-7); δ-BHC (319-86-8); γ-BHC (Lindane) (58-89-9); 4,4'-DDD (72-54-8); 4,4'-DDE (72-55-9); 4,4'-DDT (50-29-3); Dieldrin (60-57-1); Endosulfan I (959-98-8); Endosulfan II (33213-65-9); Endosulfan sulfate (1031-07-8); Endrin (72-20-8); Endrin aldehyde (7421-93-4); Endrin ketone (53494-70-5); Heptachlor (76-44-8); Heptachlor epoxide (isomer B) (1024-57-3); Methoxychlor (72-43-5)

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.