SV Calibration Mix #5/610 PAH Mix (16 Components), Restek
Catalog # RK31011
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:SV Calibration Mix #5/610 PAH Mix
- Size:1 ml/Ampule
- Cat. no.:RK31011
- Certification:CRM
- Supplier No.:31011
Specifications
About this item
Contains: Acenaphthene (83-32-9); Acenaphthylene (208-96-8); Anthracene (120-12-7); Benz(a)anthracene (56-55-3); Benzo(a)pyrene (50-32-8); Benzo(b)fluoranthene (205-99-2); Benzo(k)fluoranthene (207-08-9); Benzo(ghi)perylene (191-24-2); Chrysene (218-01-9); Dibenz(a,h)anthracene (53-70-3); Fluoranthene (206-44-0); Fluorene (86-73-7); Indeno(1,2,3-cd)pyrene (193-39-5); Naphthalene (91-20-3); Phenanthrene (85-01-8); Pyrene (129-00-0)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.