Order Entry
Canada
ContactUsLinkComponent
SV Calibration Mix #3 (14 Components), Restek
SV Calibration Mix #3 (14 Components), Restek
Catalog # RK31009
Supplier:  Restek
CAS Number:  
undefined
SV Calibration Mix #3 (14 Components), Restek
Catalog # RK31009
Supplier:  Restek
Supplier Number:  31009
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Description:
    SV Calibration Mix #3
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK31009
  • Certification:
    CRM
  • Supplier No.:
    31009

Specifications

About this item

Contains: Bis(2-chloroethoxy)methane (111-91-1); Bis(2-chloroethyl)ether (111-44-4); Bis(2-ethylhexyl)phthalate (117-81-7); 4-Bromophenyl phenyl ether (101-55-3); Benzyl butyl phthalate (85-68-7); 2-Chloronaphthalene (91-58-7); 4-Chlorophenyl phenyl ether (7005-72-3); Dimethylphthalate (131-11-3); Di-n-butylphthalate (84-74-2); Di-n-octylphthalate (117-84-0); N-Nitrosodimethylamine (62-75-9); N-Nitroso-di-n-propylamine (621-64-7); N-Nitrosodiphenylamine (86-30-6); 2,2'-Oxybis(1-chloropropane) (108-60-1)

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.