Order Entry
Canada
ContactUsLinkComponent
CLP 04.1 VOA CAL2000 MegaMix Standard (40 components), Restek
CLP 04.1 VOA CAL2000 MegaMix Standard (40 components), Restek
Catalog # RK30456
Supplier:  Restek
CAS Number:  
undefined
CLP 04.1 VOA CAL2000 MegaMix Standard (40 components), Restek
Catalog # RK30456
Supplier:  Restek
Supplier Number:  30456
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Description:
    CLP 04.1 VOA CAL2000 MegaMix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK30456
  • Certification:
    CRM
  • Supplier No.:
    30456

Specifications

About this item

Contains: Benzene (71-43-2); Bromodichloromethane (75-27-4); Bromoform (75-25-2); Carbon disulfide (75-15-0); Carbon tetrachloride (56-23-5); Chlorobenzene (108-90-7); Chloroform (67-66-3); Cyclohexane (110-82-7); Dibromochloromethane (124-48-1); 1,2-Dibromo-3-chloropropane (DBCP) (96-12-8); 1,2-Dibromoethane (EDB) (106-93-4); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); 1,1-Dichloroethane (75-34-3); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); cis-1,2-Dichloroethene (156-59-2); trans-1,2-Dichloroethene (156-60-5); 1,2-Dichloropropane (78-87-5); cis-1,3-Dichloropropene (10061-01-5); trans-1,3-Dichloropropene (10061-02-6); Ethylbenzene (100-41-4); Isopropylbenzene (cumene) (98-82-8); Methyl acetate (79-20-9); Methyl tert-butyl ether (MTBE) (1634-04-4); Methylcyclohexane (108-87-2); Methylene chloride (dichloromethane) (75-09-2); Styrene (100-42-5); 1,1,2,2-Tetrachloroethane (79-34-5); Tetrachloroethene (127-18-4); Toluene (108-88-3); 1,2,4-Trichlorobenzene (120-82-1); 1,1,1-Trichloroethane (71-55-6); 1,1,2-Trichloroethane (79-00-5); Trichloroethene (79-01-6); 1,1,2-Trichloro-1,2,2-trifluoroethane (CFC-113) (76-13-1); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.