502.2 Calibration Mix #1 (gases) (6 components), Restek
Catalog # RK30439
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in P and T methanol
- Description:502.2 Calibration Mix #1A
- Size:1 ml/Ampule
- Cat. no.:RK30439
- Certification:CRM
- Supplier No.:30439
Specifications
About this item
Contains: Bromomethane (methyl bromide) (74-83-9); Chloroethane (ethyl chloride) (75-00-3); Chloromethane (methyl chloride) (74-87-3); Dichlorodifluoromethane (CFC-12) (75-71-8); Trichlorofluoromethane (CFC-11) (75-69-4); Vinyl chloride (75-01-4)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.