MA Volatile Petroleum Hydrocarbon (VPH) Standard (13 components), Restek
Catalog # RK30434
Supplier: Restek
CAS Number:
Specifications
- Concentration:In P and T methanol
- Description:MA Volatile Petroleum Hydrocarbon (VPH) Standard
- Size:1 ml/Ampule
- Cat. no.:RK30434
- Certification:CRM
- Supplier No.:30434
Specifications
About this item
Contains: (C5) n-Pentane (109-66-0), 1,000 µg/mL; (C9) n-Nonane (111-84-2), 1,000 µg/mL; Benzene (71-43-2), 500 µg/mL; Ethylbenzene (100-41-4), 500 µg/mL; 2-Methylpentane (107-83-5), 1,500 µg/mL; Methyl tert-butyl ether (MTBE) (1634-04-4), 1,500 µg/mL; Naphthalene (91-20-3), 1,000 µg/mL; Toluene (108-88-3), 1,500 µg/mL; 1,2,4-Trimethylbenzene (95-63-6), 1,000 µg/mL; 2,2,4-Trimethylpentane (isooctane) (540-84-1), 1,500 µg/mL; m-Xylene (108-38-3), 1,000 µg/mL; o-Xylene (95-47-6), 1,000 µg/mL; p-Xylene (106-42-3), 1,000 µg/mL
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.