8240/8260 Calibration Check Mix (6 components), Restek
Catalog # RK30427
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Description:8240/8260 Calibration Check Mix
- Size:1 ml/Ampule
- Cat. no.:RK30427
- Certification:CRM
- Supplier No.:30427
Specifications
About this item
Contains: Chloroform (67-66-3); 1,1-Dichloroethene (75-35-4); 1,2-Dichloropropane (78-87-5); Ethylbenzene (100-41-4); Toluene (108-88-3); Vinyl chloride (75-01-4)