1,3,5-Trimethylbenzene, Restek
Catalog # RK30423
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:108-67-8
- Description:1,3,5-Trimethylbenzene
- Size:1 ml/Ampule
- Formula:C₉H₁₂
- Description:1,3,5-Trimethylbenzene
- Cat. no.:RK30423
- Certification:CRM
- Supplier No.:30423
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.