1,1,2,2-Tetrachloroethane, Restek
Catalog # RK30412
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:79-34-5
- Description:1,1,2,2-Tetrachloroethane
- Size:1 ml/Ampule
- Formula:C₂H₂Cl₄
- Description:1,1,2,2-Tetrachloroethane
- Cat. no.:RK30412
- Certification:CRM
- Supplier No.:30412
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.