Order Entry
Canada
ContactUsLinkComponent
Dibromochloromethane (chlorodibromochloromethane), Restek
Dibromochloromethane (chlorodibromochloromethane), Restek
Catalog # RK30271
Supplier:  Restek
CAS Number:  
undefined
Dibromochloromethane (chlorodibromochloromethane), Restek
Catalog # RK30271
Supplier:  Restek
Supplier Number:  30271
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in P and T methanol
  • CAS No.:
    124-48-1
  • Description:
    Dibromochloromethane (chlorodibromochloromethane)
  • Size:
    1 ml/Ampule
  • Formula:
    CHBr₂Cl
  • Description:
    Dibromochloromethane (chlorodibromochloromethane)
  • Cat. no.:
    RK30271
  • Certification:
    CRM
  • Supplier No.:
    30271

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.