Order Entry
Canada
ContactUsLinkComponent
8020A Calibration Mix (10 components), Restek
8020A Calibration Mix (10 components), Restek
Catalog # RK30222
Supplier:  Restek
CAS Number:  
undefined
8020A Calibration Mix (10 components), Restek
Catalog # RK30222
Supplier:  Restek
Supplier Number:  30222
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Description:
    8020A Calibration Mix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK30222
  • Certification:
    CRM
  • Supplier No.:
    30222

Specifications

About this item

Mix contains: Benzene (71-43-2); Chlorobenzene (108-90-7); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); Ethylbenzene (100-41-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.