Order Entry
Canada
ContactUsLinkComponent
PVOC/GRO Mix (Wisconsin) (10 components), Restek
PVOC/GRO Mix (Wisconsin) (10 components), Restek
Catalog # RK30095
Supplier:  Restek
CAS Number:  
undefined
PVOC/GRO Mix (Wisconsin) (10 components), Restek
Catalog # RK30095
Supplier:  Restek
Supplier Number:  30095
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in P and T methanol
  • Description:
    PVOC/GRO Mix (Wisconsin)
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK30095
  • Certification:
    CRM
  • Supplier No.:
    30095

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.