Order Entry
Canada
ContactUsLinkComponent
8011 Calibration Mix—EDB/DBCP (2 components), Restek
8011 Calibration Mix—EDB/DBCP (2 components), Restek
Catalog # RK30062
Supplier:  Restek
CAS Number:  
undefined
8011 Calibration Mix—EDB/DBCP (2 components), Restek
Catalog # RK30062
Supplier:  Restek
Supplier Number:  30062
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Description:
    8011 Calibration Mix—EDB/DBCP
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK30062
  • Certification:
    CRM
  • Supplier No.:
    30062

Specifications

About this item

Mix contains: 1,2-Dibromo-3-chloropropane (DBCP) (96-12-8), 1,2-Dibromoethane (EDB) (106-93-4).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.