Order Entry
Canada
ContactUsLinkComponent
DW-VOC Mix #1 (8 components), Restek
DW-VOC Mix #1 (8 components), Restek
Catalog # RK30219
Supplier:  Restek
CAS Number:  
undefined
DW-VOC Mix #1 (8 components), Restek
Catalog # RK30219
Supplier:  Restek
Supplier Number:  30219
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Description:
    DW-VOC Mix #1A
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK30219
  • Certification:
    CRM
  • Supplier No.:
    30219

Specifications

About this item

Contains: Benzene (71-43-2); Carbon tetrachloride (56-23-5); 1,4-Dichlorobenzene (106-46-7); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); 1,1,1-Trichloroethane (71-55-6); Trichloroethene (79-01-6); Vinyl chloride (75-01-4)