602 Purgeable Aromatics Calibration Mix (7 components), Restek
RK30035
: Restek
:
- Concentration:2000 µg/ml each in P and T methanol
- Description:602 Purgeable Aromatics Calibration Mix
- Size:1 ml/Ampule
- Cat. no.:RK30035
- Certification:CRM
- Supplier No.:30035
Mix contains: Benzene (71-43-2), Chlorobenzene (108-90-7), 1,2-Dichlorobenzene (95-50-1), 1,3-Dichlorobenzene (541-73-1), 1,4-Dichlorobenzene (106-46-7), Ethylbenzene (100-41-4), Toluene (108-88-3).
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.